Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM7SF3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen TM7SF3
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123366
SDP
View Documents
Novus Biologicals
NBP30923225UL
25 μg
Each for $206.00
Only null left
Add to Cart
 
NB123365
SDP
View Documents
Novus Biologicals
NBP309232100UL
100 μg
Each for $487.50
Only null left
Add to Cart
 
Description

Description

TM7SF3 Polyclonal specifically detects TM7SF3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TM7SF3
Western Blot
Unconjugated
Rabbit
Human
seven span transmembrane protein, seven transmembrane protein TM7SF3, transmembrane 7 superfamily member 3
The immunogen is a synthetic peptide directed towards the middle region of Human TM7S3. Peptide sequence LLKHLQRMVSVPQVKASALKVVTLTANDKTSVSFSSLPGQGVIYNVIVWD
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
51768
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.