Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TM7SF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589951
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589951 Supplier Invitrogen™ Supplier No. PA589951
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KILVSASFYP SVERKRIQYL HAKLLKKRSK QPLGEVKRRL SLYLTKIHFW LPVLKMIRKK QMDMASADKS; Positive Samples: 293T; Cellular Location: Cell membrane, Endoplasmic reticulum membrane, Endoplasmic reticulum-Golgi intermediate compartment membrane, Endosome, Multi-pass membrane protein.

DCSTAMP gene ontology annotations related to this gene include cellular response to interleukin-4; cellular response to macrophage colony-stimulating factor stimulus; cellular response to tumor necrosis factor; membrane fusion; myeloid dendritic cell differentiation; negative regulation of cell growth; osteoclast fusion; positive regulation of bone resorption; positive regulation of macrophage fusion; positive regulation of monocyte differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TM7SF4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DCSTAMP
Gene Accession No. Q7TNJ0, Q9H295
Gene Alias 4833414I07Rik; DC-specific transmembrane protein; Dcstamp; DC-STAMP; Dendritic cells (DC)-specific transmembrane protein; Dendritic cell-specific transmembrane protein; dendrocyte expressed seven transmembrane protein; dendrocyte-expressed seven transmembrane protein; dentrocyte expressed seven transmembrane protein; FIND; hDC-STAMP; IL-4-induced protein; IL-Four (IL-4) induced; IL-Four INDuced; IL-four-induced protein; LOW QUALITY PROTEIN: dendritic cell-specific transmembrane protein; mDC-STAMP; TM7SF4; transmembrane 7 superfamily member 4
Gene Symbols DCSTAMP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 401-470 of human DCSTAMP (NP_1104151).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75766, 81501
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.