Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TM9SF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542621 Supplier Invitrogen™ Supplier No. PA542621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EGVTHYKAGD PVILYVNKVG PYHNPQETYH YYQLPVCCPE KIRHKSLSLG Sequence homology: Cow: 100%; Dog: 100%; Goat: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Plays an essential role in autophagy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TM9SF1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TM9SF1
Gene Accession No. O15321
Gene Alias 1200014D02Rik; AI893436; fa03b09; HMP70; MP70; MP70 protein family member; multispanning membrane protein (70kD); TM9SF1; Transmembrane 9 superfamily member 1; transmembrane protein 9 superfamily member 1; wu:fa03b09; zgc:100810
Gene Symbols TM9SF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TM9SF1 (aa 31-80).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10548
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.