Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TM9SF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542622
Description
Peptide sequence: THTYSVRWSE TSVERRSDRR RGDDGGFFPR TLEIHWLSII NSMVLVFLLV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Plays an essential role in autophagy.
Specifications
TM9SF1 | |
Polyclonal | |
Unconjugated | |
TM9SF1 | |
1200014D02Rik; AI893436; fa03b09; HMP70; MP70; MP70 protein family member; multispanning membrane protein (70kD); TM9SF1; Transmembrane 9 superfamily member 1; transmembrane protein 9 superfamily member 1; wu:fa03b09; zgc:100810 | |
Rabbit | |
Protein A | |
RUO | |
10548 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O15321 | |
TM9SF1 | |
Synthetic peptide directed towards the middle region of human TM9SF1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction