Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TM9SF1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS10764R

Description
Plays an essential role in autophagy.Specifications
TM9SF1 | |
Polyclonal | |
Unconjugated | |
TM9SF1 | |
1200014D02Rik; AI893436; BOS_10209; fa03b09; hMP70; importin 4; IPO4; MP70; MP70 protein family member; multispanning membrane protein (70kD); TM9SF1; Transmembrane 9 superfamily member 1; transmembrane protein 9 superfamily member 1; wu:fa03b09; zgc:100810 | |
Rabbit | |
Protein A | |
RUO | |
10548 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH | |
O15321 | |
TM9SF1 | |
KLH conjugated synthetic peptide derived from human TM9SF1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction