Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TM9SF3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558855
Description
Immunogen sequence: SLPFCVGSKK SISHYHETLG EALQGVELEF SGLDIKFKDD VMPATYCEID LDKEKRDAFV YAIKNHYWYQ MYIDDLPIWG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
TM9SF3 is a protein coding gene.
Specifications
TM9SF3 | |
Polyclonal | |
Unconjugated | |
TM9SF3 | |
1810073M23Rik; 2810031D16Rik; AI115521; AI413748; AW146116; AW549777; dinucleotide oxidase disulfide thiol exchanger 3 superfamily member 3; endomembrane protein emp70 precursor isolog; EP70-P-iso; fb75h12; hypothetical protein LOC406698; mKIAA4036; P70-P-iso; RGD1564625; RP11-34E5.1; SM-11044 binding protein; SM-11044-binding protein; SMBP; Tm9sf3; Transmembrane 9 superfamily member 3; Transmembrane 9 superfamily member 3-like protein; transmembrane protein 9 superfamily member 3; UNQ245/PRO282; wu:fa94f05; wu:fb75h12; zgc:56246 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56889 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HD45 | |
TM9SF3 | |
Recombinant protein corresponding to Human TM9SF3. Recombinant protein control fragment (Product #RP-97012). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction