Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TM9SF3 Polyclonal Antibody
TM9SF3 Polyclonal Antibody
Supplier: Thermo Scientific PA569606
Description
TM9SF3 Polyclonal Antibody for Western Blot

Specifications
TM9SF3 | |
Polyclonal | |
Unconjugated | |
TM9SF3 | |
1810073M23Rik; 2810031D16Rik; AI115521; AI413748; AW146116; AW549777; dinucleotide oxidase disulfide thiol exchanger 3 superfamily member 3; endomembrane protein emp70 precursor isolog; EP70-P-iso; fb75h12; hypothetical protein LOC406698; mKIAA4036; P70-P-iso; RGD1564625; RP11-34E5.1; SM-11044 binding protein; SM-11044-binding protein; SMBP; Tm9sf3; Transmembrane 9 superfamily member 3; Transmembrane 9 superfamily member 3-like protein; transmembrane protein 9 superfamily member 3; UNQ245/PRO282; wu:fa94f05; wu:fb75h12; zgc:56246 | |
Rabbit | |
Affinity chromatography | |
RUO | |
309475 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
TM9SF3 | |
synthetic peptide directed towards the N-terminal region of rat TM9SF3. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction