Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TM9SF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534268
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534268 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534268 Supplier Invitrogen™ Supplier No. PA534268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Galago, Marmoset, Mouse, Rabbit, Guinea pig (100%).

TM9SF4 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TM9SF4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TM9SF4
Gene Accession No. Q8BH24, Q92544
Gene Alias AA986553; AU045326; B930079E06; dinucleotide oxidase disulfide thiol exchanger 3 superfamily member 4; dJ836N17.2; Kiaa0255; LOW QUALITY PROTEIN: transmembrane 9 superfamily member 4; mKIAA0255; TM9SF4; Transmembrane 9 superfamily member 4; transmembrane 9 superfamily protein member 4; TUCAP1; Tumor cannibalism associated protein 1; zgc:66234
Gene Symbols TM9SF4
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from internal region of human TM9SF4.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100343710, 100733724, 9777, 99237
Target Species Human, Mouse, Rabbit, Monkey, Guinea Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.