Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TM9SF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584627
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584627 Supplier Invitrogen™ Supplier No. PA584627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDYYVHLIAD NLPVATRLEL YSNRDSDDKK KEKDVQFEHG YRLGFTDVNK IYLHNHLSFI LYYHREDMEE DQEHTYRVVR FE Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 98%.

TM9SF4 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TM9SF4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TM9SF4
Gene Accession No. Q92544
Gene Alias AA986553; AU045326; B930079E06; dinucleotide oxidase disulfide thiol exchanger 3 superfamily member 4; dJ836N17.2; Kiaa0255; LOW QUALITY PROTEIN: transmembrane 9 superfamily member 4; mKIAA0255; TM9SF4; Transmembrane 9 superfamily member 4; transmembrane 9 superfamily protein member 4; TUCAP1; Tumor cannibalism associated protein 1; zgc:66234
Gene Symbols TM9SF4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TM9SF4. Recombinant protein control fragment (Product #RP-99324).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9777
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.