Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TM9SF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569446
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
TM9SF4 is a protein coding gene.
Specifications
| TM9SF4 | |
| Polyclonal | |
| Unconjugated | |
| TM9SF4 | |
| AA986553; AU045326; B930079E06; dinucleotide oxidase disulfide thiol exchanger 3 superfamily member 4; dJ836N17.2; Kiaa0255; LOW QUALITY PROTEIN: transmembrane 9 superfamily member 4; mKIAA0255; TM9SF4; Transmembrane 9 superfamily member 4; transmembrane 9 superfamily protein member 4; TUCAP1; Tumor cannibalism associated protein 1; zgc:66234 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9777 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q92544 | |
| TM9SF4 | |
| synthetic peptide directed towards the N terminal region of human TM9SF4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction