Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMA16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111360
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111360 Supplier Invitrogen™ Supplier No. PA5111360
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KALRLNLVGE KLQWFQNHLD PQKKRYSKKD ACELIERYLN RFSSELEQIE LHNSIRDRQG RRHCSRETVI KQTMER Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 82%.

TMA16 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMA16
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMA16
Gene Accession No. Q96EY4
Gene Alias 1810029B16Rik; C4orf43; RGD1305222; TMA16; translation machinery associated 16 homolog; translation machinery associated 16 homolog (S. cerevisiae); translation machinery-associated protein 16; UPF0534 protein C4orf43; UPF0534 protein C4orf43 homolog
Gene Symbols TMA16
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TMA16. Recombinant protein control fragment (Product #RP-106410).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55319
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.