Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMA7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561500
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561500 Supplier Invitrogen™ Supplier No. PA561500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSEGGKKKPL KQPKKQAKEM DEEDKAFKQK QKEEPKKLE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

CCDC72, also named as TMA7, is a novel modulator for hair follicle development. CCDC72 has the promoter activity and regulation of gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMA7
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tma7
Gene Accession No. Q9Y2S6
Gene Alias 1110017O22Rik; CCDC72; coiled-coil domain containing 72; coiled-coil domain-containing protein 72; HSPC016; HSPC330; Tma7; translation machinery associated 7 homolog; translation machinery-associated protein 7; translational machinery associated 7 homolog; translational machinery associated 7 homolog (S. cerevisiae)
Gene Symbols Tma7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMA7. Recombinant protein control fragment (Product #RP-100566).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51372
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.