Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMBIM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551044
Description
TMBIM1 Polyclonal Antibody detects endogenous levels of TMBIM1 protein.
Negatively regulates aortic matrix metalloproteinase-9 (MMP9) production and may play a protective role in vascular remodeling.
Specifications
TMBIM1 | |
Polyclonal | |
Unconjugated | |
TMBIM1 | |
2310061B02Rik; AA960455; AU024746; C78899; Lfg3; mKIAA4161; MST100; MSTP100; PP1201; Protein lifeguard 3; Protein RECS1; Protein RECS1 homolog; PSEC0158; RECS1; Responsive to centrifugal force and shear stress gene 1 protein; Tmbib1; Tmbim1; transmembrane BAX inhibitor motif containing 1; transmembrane BAX inhibitor motif-containing protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
316516, 64114, 69660 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8BJZ3, Q969X1 | |
TMBIM1 | |
Synthesized peptide derived from the C-terminal region of human RECS1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction