Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMBIM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545924
Description
Peptide sequence: LTVAVVVTFY DVYIILQAFI LTTTVFFGLT VYTLQSKKDF SKFGAGLFAL Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 93%.
TMBIM4 is an anti-apoptotic protein which can inhibit apoptosis induced by intrinsic and extrinsic apoptotic stimuli. It can modulate both capacitative Ca2+ entry and inositol 1, 4, 5-trisphosphate (IP3)-mediated Ca2+ release.
Specifications
| TMBIM4 | |
| Polyclonal | |
| Unconjugated | |
| TMBIM4 | |
| 0610007H07Rik; AU022431; Cgi119; CGI-119; CGI-119 protein; GAAP; Golgi anti-apoptotic protein; hypothetical protein LOC513242; LFG4; Protein lifeguard 4; Protein S1R; S1R; Tmbim4; transmembrane BAX inhibitor motif containing 4; transmembrane BAX inhibitor motif-containing protein 4; transmembrane BAX inhibitor motif-containing protein 4; protein lifeguard 4; ZPRO; Z-protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51643 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9HC24 | |
| TMBIM4 | |
| Synthetic peptide directed towards the middle region of human TMBIM4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction