Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMBIM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567464 Supplier Invitrogen™ Supplier No. PA567464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RYPRSSIEDDF NYGSSVASAT VHIRMVLQRD CTFSKQYMRI PSAPSATLNI IRFFHLRQSG RSGMVNIFYK GPTFL Highest antigen sequence identity to the following orthologs - mouse 32%, rat 33%.

TMBIM4 is an anti-apoptotic protein which can inhibit apoptosis induced by intrinsic and extrinsic apoptotic stimuli. It can modulate both capacitative Ca2+ entry and inositol 1, 4, 5-trisphosphate (IP3)-mediated Ca2+ release.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMBIM4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMBIM4
Gene Accession No. Q9HC24
Gene Alias 0610007H07Rik; AU022431; Cgi119; CGI-119; CGI-119 protein; GAAP; Golgi anti-apoptotic protein; hypothetical protein LOC513242; LFG4; Protein lifeguard 4; Protein S1R; S1R; Tmbim4; transmembrane BAX inhibitor motif containing 4; transmembrane BAX inhibitor motif-containing protein 4; transmembrane BAX inhibitor motif-containing protein 4; protein lifeguard 4; ZPRO; Z-protein
Gene Symbols TMBIM4
Host Species Rabbit
Immunogen Recombinant Human TMBIM4. Recombinant protein control fragment (Product #RP-108119).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51643
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.