Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMBIM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104271
Description
Antibody detects endogenous levels of total GAAP.
TMBIM4 is an anti-apoptotic protein which can inhibit apoptosis induced by intrinsic and extrinsic apoptotic stimuli. It can modulate both capacitative Ca2+ entry and inositol 1, 4, 5-trisphosphate (IP3)-mediated Ca2+ release.
Specifications
TMBIM4 | |
Polyclonal | |
Unconjugated | |
TMBIM4 | |
0610007H07Rik; AU022431; Cgi119; CGI-119; CGI-119 protein; GAAP; Golgi anti-apoptotic protein; hypothetical protein LOC513242; LFG4; Protein lifeguard 4; Protein S1R; S1R; Tmbim4; transmembrane BAX inhibitor motif containing 4; transmembrane BAX inhibitor motif-containing protein 4; transmembrane BAX inhibitor motif-containing protein 4; protein lifeguard 4; ZPRO; Z-protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
51643 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9HC24 | |
TMBIM4 | |
A synthesized peptide derived from human TMBIM4(Accession Q9HC24), corresponding to amino acid residues M1-K37. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction