Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561222
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561222 Supplier Invitrogen™ Supplier No. PA561222
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QVLREVEKSH KSVKGKATAR DSEDTPKSSS KNATQLQLTK EETTPPSASQ SQAMDKKAQG PGTSNSASRT Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 61%.

This gene is considered a member of a gene family predicted to encode transmembrane proteins. The specific function of this gene is unknown; however, expression in the inner ear suggests that it may be crucial for normal auditory function. Mutations in this gene may underlie hereditary disorders of balance and hearing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMC2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMC2
Gene Accession No. Q8TDI7
Gene Alias 4933416G09Rik; Beethoven; Bth; C20orf145; CWEA1; CWEA2; dJ686C3.3; dn; TMC2; transmembrane channel like 2; transmembrane channel-like 2; transmembrane channel-like gene family 2; transmembrane channel-like protein 2; transmembrane cochlear-expressed gene 2; transmembrane cochlear-expressed protein 2; transmembrane, cochlear expressed 2; UNQ907/PRO1928
Gene Symbols TMC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMC2. Recombinant protein control fragment (Product #RP-99443).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117532
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.