Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561788
Description
Immunogen sequence: EEDGGRSRKA FTEVTQTELQ DPHPSRELPW PMQARRAHRQ RNASRDQVVY GSGTKTDRWA RLLRRSKEKT KEGLRSLQPW AWTLK Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 56%.
Probable ion channel. [UniProt].
Specifications
TMC4 | |
Polyclonal | |
Unconjugated | |
TMC4 | |
testicular tissue protein Li 204; TMC4; transmembrane channel like 4; transmembrane channel-like 4; transmembrane channel-like gene family 4; transmembrane channel-like protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
147798 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z404 | |
TMC4 | |
Recombinant protein corresponding to Human TMC4. Recombinant protein control fragment (Product #RP-99148). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction