Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMC5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559308
Description
Immunogen sequence: TEGRKIMIRL LHEQIINEGK DKMFLIEKLI KLQDMEKKAN PSSLVLERRE VEQQGFLHLG EHDGSLDLRS RRSVQEGNPR Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.
May function as ion channels, transporters, or modulators of such. Ubiquitously expressed.
Specifications
TMC5 | |
Polyclonal | |
Unconjugated | |
TMC5 | |
4932443L08Rik; AU016085; Tmc5; transmembrane channel like 5; transmembrane channel-like 5; transmembrane channel-like gene family 5; transmembrane channel-like protein 5; UNQ8238/PRO33604 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79838 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UXY8 | |
TMC5 | |
Recombinant protein corresponding to Human TMC5. Recombinant protein control fragment (Product #RP-98636). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction