Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMC5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559834
Description
Immunogen sequence: STWREPDYSD AENGHDYGSS ETPKMTRGVL SRTSSIQPSF RHRSDDPVGS LWGENDYPEG IEMASMEMAN SYGHSLPGAP GSGYVNPAYV GESGPVH Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 42%.
May function as ion channels, transporters, or modulators of such. Ubiquitously expressed.
Specifications
TMC5 | |
Polyclonal | |
Unconjugated | |
TMC5 | |
4932443L08Rik; AU016085; Tmc5; transmembrane channel like 5; transmembrane channel-like 5; transmembrane channel-like gene family 5; transmembrane channel-like protein 5; UNQ8238/PRO33604 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79838 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UXY8 | |
TMC5 | |
Recombinant protein corresponding to Human TMC5. Recombinant protein control fragment (Product #RP-98635). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction