Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595696
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595696 Supplier Invitrogen™ Supplier No. PA595696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YWQITEGRKI MIRLLHEQII NEGKDKMFLI EKLIKLQDME KKANPSSLVL ERREVEQQGF LHLGEHDGSL DLRSRRSVQE GNPRA; Positive Samples: BT-474; Cellular Location: Membrane, Multi-pass membrane protein.

May function as ion channels, transporters, or modulators of such. Ubiquitously expressed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMC5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TMC5
Gene Accession No. Q6UXY8
Gene Alias 4932443L08Rik; AU016085; Tmc5; transmembrane channel like 5; transmembrane channel-like 5; transmembrane channel-like gene family 5; transmembrane channel-like protein 5; UNQ8238/PRO33604
Gene Symbols TMC5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 676-760 of human TMC5 (NP_0790562).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79838
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.