Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMC7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104440
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104440 Supplier Invitrogen™ Supplier No. PA5104440
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TMC7.

TMC7 (Transmembrane channel-like protein 7) is a 723 amino acid protein that is a member of the TMC protein family. All TMC genes encode transmembrane proteins with intracellular amino- and carboxy- termini and at least eight membrane spanning domains. Therefore, TMC7 is a multi-pass membrane protein that may regulate or function as an ion channel or transporter. The gene encoding TMC7 maps to human chromosome 16, which encodes over 900 genes and comprises nearly 3% of the human genome. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16, as is Crohn's disease, which is a gastrointestinal inflammatory condition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMC7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TMC7
Gene Accession No. Q7Z402, Q8C428
Gene Alias 1700030H01Rik; C230064B05; C630024K23Rik; RGD1564267; Tmc7; transmembrane channel like 7; transmembrane channel-like 7; transmembrane channel-like gene family 7; transmembrane channel-like protein 7
Gene Symbols TMC7
Host Species Rabbit
Immunogen A synthesized peptide derived from human TMC7(Accession Q7Z402), corresponding to amino acid residues C676-N723.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209760, 499254, 79905
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.