Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542627
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542627 Supplier Invitrogen™ Supplier No. PA542627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ERLEEQLNDL TELHQNEILN LKQELASMEE KIAYQSYERA RDIQEALEAC Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

TMCC1 is a multi-pass membrane protein. It belongs to the TEX28 family. The exact function of TMCC1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMCC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMCC1
Gene Accession No. O94876
Gene Alias 3632431M01Rik; Kiaa0779; mKIAA0779; RGD1311909; TMCC1; transmembrane and coiled coil domains 1; transmembrane and coiled-coil domain family 1; transmembrane and coiled-coil domains protein 1
Gene Symbols TMCC1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TMCC1 (aa 506-555).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23023
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.