Learn More
Invitrogen™ TMCC2 Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100% Immunogen sequence: KVDKGDLVAL SLPAGHGDTD GPISLDVPDG APDPQRTKAA IDHLHQKILK.
Specifications
Specifications
| Antigen | TMCC2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | TMCC2 |
| Gene Accession No. | O75069 |
| Gene Alias | 1110063G11Rik; cerebral protein 11; HUCEP11; hucep-11; Kiaa0481; RGD1311960; TMCC2; transmembrane and coiled-coil domain family 2; transmembrane and coiled-coil domains 2; transmembrane and coiled-coil domains protein 2 |
| Gene Symbols | TMCC2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.