Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMCO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543350
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543350 Supplier Invitrogen™ Supplier No. PA543350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CSFIFLYILC TMSIRQNIQK ILGLAPSRAA TKQAGGFLGP PPPSGKFS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

TMCO1 belongs to the TMCO1 family. The exact function of TMCO1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMCO1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMCO1
Gene Accession No. Q9UM00
Gene Alias 1190006A08Rik; 4930403O06Rik; AA109065; AU022572; calcium load-activated calcium channel; CLAC channel; ESTM39; HP10122; hypothetical protein LOC614715; LOW QUALITY PROTEIN: transmembrane and coiled-coil domain-containing protein 1; meg-2-like protein; PCIA3; PNAS-10; PNAS-136; putative membrane protein; sb:cb729; TMCC4; tmco1; transmembrane and coiled-coil domain-containing 1-like protein; transmembrane and coiled-coil domain-containing protein 1; transmembrane and coiled-coil domains 1; transmembrane and coiled-coil domains 4; transmembrane and coiled-coil domains protein 1; transmembrane and coiled-coil domains protein 1-like; Transmembrane and coiled-coil domains protein 4; UNQ151/PRO177; Xenogeneic cross-immune protein PCIA3; zgc:110322; zgc:110322 protein; zgc:92740
Gene Symbols TMCO1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TMCO1 (aa 192-239).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54499
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.