Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMCO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531741 Supplier Invitrogen™ Supplier No. PA531741
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431. Predicted reactivity: Mouse (96%), Rat (95%), Pig (90%), Chicken (95%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Probable Na(+)/H(+) antiporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMCO3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene TMCO3
Gene Accession No. Q6UWJ1
Gene Alias B230339H12Rik; C13orf11; C87304; LOW QUALITY PROTEIN: transmembrane and coiled-coil domain-containing protein 3; putative LAG1-interacting protein; RGD1306586; testis tissue sperm-binding protein Li 36a; tmco3; Transmembrane and coiled-coil domain-containing protein 3; transmembrane and coiled-coil domain-containing protein 3; LOW QUALITY PROTEIN: transmembrane and coiled-coil domain-containing protein 3; transmembrane and coiled-coil domains 3; UNQ2419/PRO4976
Gene Symbols TMCO3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TMCO3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55002
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.