Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMCO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558839 Supplier Thermo Scientific Supplier No. PA558839
Add to Cart
Add to Cart

TMCO3 Polyclonal Antibody

TMCO3 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMCO3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMCO3
Gene Accession No. Q6UWJ1
Gene Alias B230339H12Rik; C13orf11; C87304; LOW QUALITY PROTEIN: transmembrane and coiled-coil domain-containing protein 3; putative LAG1-interacting protein; RGD1306586; testis tissue sperm-binding protein Li 36a; tmco3; Transmembrane and coiled-coil domain-containing protein 3; transmembrane and coiled-coil domain-containing protein 3; LOW QUALITY PROTEIN: transmembrane and coiled-coil domain-containing protein 3; transmembrane and coiled-coil domains 3; UNQ2419/PRO4976
Gene Symbols TMCO3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMCO3. Recombinant protein control fragment (Product #RP-95653).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55002
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.