Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMCO5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113222
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113222 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113222 Supplier Invitrogen™ Supplier No. PA5113222
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMCO5A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMCO5A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TMCO5A
Gene Accession No. Q8N6Q1
Gene Alias testicular tissue protein Li 205; TMCO5; TMCO5A; Transmembrane and coiled-coil domain-containing protein 5A; transmembrane and coiled-coil domains 5; transmembrane and coiled-coil domains 5A
Gene Symbols TMCO5A
Host Species Rabbit
Immunogen Recombinant Human Transmembrane and coiled-coil domain-containing protein 5A protein (10-192AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145942
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.