Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMCO5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571528
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571528 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571528 Supplier Invitrogen™ Supplier No. PA571528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMCO5B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMCO5B
Applications Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TMCO5B
Gene Accession No. A8MYB1
Gene Alias TMCO5B; transmembrane and coiled-coil domains 5B (pseudogene)
Gene Symbols TMCO5B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 210-238 amino acids from the C-terminal region of human TMCO5B.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100652857
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.