Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMED1 Antibody (1009527), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2243Y
Description
TMED1 Monoclonal antibody specifically detects TMED1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
TMED1 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
11018 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, Immunocytochemistry, CyTOF | |
1009527 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
Il1rl1l, IL1RL1LGIL1RL1-binding protein, interleukin 1 receptor-like 1 ligand, Interleukin-1 receptor-like 1 ligand, MGC1270, Putative T1/ST2 receptor-binding protein, ST2L, T1/ST2 receptor binding protein, transmembrane emp24 domain containing 1, transmembrane emp24 domain-containing protein 1, transmembrane emp24 protein transport domain containing 1 | |
Human embryonic kidney cell, HEK293 derived human TMED1, Met1-Asn194, Accession # Q13445 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction