Learn More
Invitrogen™ TMED1 Monoclonal Antibody (OTI7D2)

Description
TMED1 Monoclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | TMED1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI7D2 |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TMED1 |
| Gene Accession No. | Q13445 |
| Gene Alias | hypothetical protein LOC538144; IL1RL1-binding protein; Il1rl1l; IL1RL1LG; interleukin 1 receptor-like 1 ligand; interleukin-1 receptor-like 1 ligand; Ly84l; lymphocyte antigen 84 ligand; MGC1270; p24 family protein gamma-1; p24g1; p24gamma1; putative T1/ST2 receptor-binding protein; St2l; Tmed1; Tp24; transmembrane emp24 domain containing 1; transmembrane emp24 domain-containing protein 1; transmembrane emp24 protein transport domain containing 1; transmembrane p24 trafficking protein 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.