Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMED1 Monoclonal Antibody (OTI7D2), TrueMAB™, OriGene
SDP

Catalog No. 502540796
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0796 30 μL
50-167-4472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0796 Supplier OriGene Supplier No. TA503620S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMED1 Monoclonal antibody specifically detects TMED1 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen TMED1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI7D2
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Ung
Gene Accession No. P13051
Gene Alias hypothetical protein LOC538144; IL1RL1-binding protein; Il1rl1l; IL1RL1LG; interleukin 1 receptor-like 1 ligand; interleukin-1 receptor-like 1 ligand; Ly84l; lymphocyte antigen 84 ligand; MGC1270; p24 family protein gamma-1; p24g1; p24gamma1; putative T1/ST2 receptor-binding protein; St2l; Tmed1; Tp24; transmembrane emp24 domain containing 1; transmembrane emp24 domain-containing protein 1; transmembrane emp24 protein transport domain containing 1; transmembrane p24 trafficking protein 1
Host Species Mouse
Immunogen Full length human recombinant protein of human UNG produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7374
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.