Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMED1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542927 Supplier Invitrogen™ Supplier No. PA542927
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FTLESPQGVL LVSESRKADG VHTVEPTEAG DYKLCFDNSF STISEKLVFF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 77%.

TMED1 was identified by its interaction with interleukin 1 receptor-like 1 (IL1RL1). This protein lacks any similarity to other interleukin 1 ligands. The functional significance of its interaction with IL1RL1 is not known. The protein encoded by this gene was identified by its interaction with interleukin 1 receptor-like 1 (IL1RL1). This protein lacks any similarity to other interleukin 1 ligands. The functional significance of its interaction with IL1RL1 is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMED1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMED1
Gene Accession No. Q13445
Gene Alias hypothetical protein LOC538144; IL1RL1-binding protein; Il1rl1l; IL1RL1LG; interleukin 1 receptor-like 1 ligand; interleukin-1 receptor-like 1 ligand; Ly84l; lymphocyte antigen 84 ligand; MGC1270; p24 family protein gamma-1; p24g1; p24gamma1; putative T1/ST2 receptor-binding protein; St2l; Tmed1; Tp24; transmembrane emp24 domain containing 1; transmembrane emp24 domain-containing protein 1; transmembrane emp24 protein transport domain containing 1; transmembrane p24 trafficking protein 1
Gene Symbols TMED1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TMED1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11018
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.