Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMED1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553826
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553826 Supplier Invitrogen™ Supplier No. PA553826
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPIQDGEFTF LLPAGRKQCF YQSAPANASL ETEYQVIGGA GLDVDFTLES PQGVLLVSES RKADG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

TMED1 was identified by its interaction with interleukin 1 receptor-like 1 (IL1RL1). This protein lacks any similarity to other interleukin 1 ligands. The functional significance of its interaction with IL1RL1 is not known. The protein encoded by this gene was identified by its interaction with interleukin 1 receptor-like 1 (IL1RL1). This protein lacks any similarity to other interleukin 1 ligands. The functional significance of its interaction with IL1RL1 is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMED1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMED1
Gene Accession No. Q13445
Gene Alias hypothetical protein LOC538144; IL1RL1-binding protein; Il1rl1l; IL1RL1LG; interleukin 1 receptor-like 1 ligand; interleukin-1 receptor-like 1 ligand; Ly84l; lymphocyte antigen 84 ligand; MGC1270; p24 family protein gamma-1; p24g1; p24gamma1; putative T1/ST2 receptor-binding protein; St2l; Tmed1; Tp24; transmembrane emp24 domain containing 1; transmembrane emp24 domain-containing protein 1; transmembrane emp24 protein transport domain containing 1; transmembrane p24 trafficking protein 1
Gene Symbols TMED1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMED1. Recombinant protein control fragment (Product #RP-90026).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11018
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.