Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMED1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597310
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597310 Supplier Invitrogen™ Supplier No. PA597310
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGPPPIQDGE FTFLLPAGRK QCFYQSAPAN ASLETEYQVI GGAGLDVDFT LESPQGVLLV SESRKADGVH TVEPTEAGDY KLCFDNSFST ISEKLVFFEL IFDSLQDDEE VEGWAEAVEP EEMLDVKMED IKESIETMRT RLERSIQMLT LLRAFEARDR NLQEGNLERV N; Positive Samples: BT-474, Mouse heart; Cellular Location: Cell membrane, Endoplasmic reticulum membrane, Endoplasmic reticulum-Golgi intermediate compartment membrane, Golgi apparatus, Single-pass type I membrane protein, cis-Golgi network membrane.

TMED1 was identified by its interaction with interleukin 1 receptor-like 1 (IL1RL1). This protein lacks any similarity to other interleukin 1 ligands. The functional significance of its interaction with IL1RL1 is not known. The protein encoded by this gene was identified by its interaction with interleukin 1 receptor-like 1 (IL1RL1). This protein lacks any similarity to other interleukin 1 ligands. The functional significance of its interaction with IL1RL1 is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMED1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TMED1
Gene Accession No. Q13445, Q3V009
Gene Alias hypothetical protein LOC538144; IL1RL1-binding protein; Il1rl1l; IL1RL1LG; interleukin 1 receptor-like 1 ligand; interleukin-1 receptor-like 1 ligand; Ly84l; lymphocyte antigen 84 ligand; MGC1270; p24 family protein gamma-1; p24g1; p24gamma1; putative T1/ST2 receptor-binding protein; St2l; Tmed1; Tp24; transmembrane emp24 domain containing 1; transmembrane emp24 domain-containing protein 1; transmembrane emp24 protein transport domain containing 1; transmembrane p24 trafficking protein 1
Gene Symbols TMED1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-194 of human TMED1 (NP_0068491).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11018, 17083
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.