Learn More
Invitrogen™ TMED2 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (95%), Xenopus laevis (100%), Chicken (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | TMED2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.92 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Tmed2 |
| Gene Accession No. | Q15363 |
| Gene Alias | 1110032D12Rik; 1810020N21Rik; coated vesicle membrane protein; COPI-coated vesicle membrane protein p24; Membrane protein p24A; p24; p24 family protein beta-1; P24A; p24b1; p24beta1; RNP21.4; RNP24; sid 394; Sid394; Sid394p; TMED2; transmembrane emp24 domain trafficking protein 2; transmembrane emp24 domain-containing protein 2; transmembrane p24 trafficking protein 2 |
| Gene Symbols | Tmed2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.