Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMED3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110494
Description
Immunogen sequence: PCGAELTFEL PDNAKQCFHE EVEQGVKFSL DYQVITGGHY DVDCYVEDPQ GNTIYRETKK QYDSFTYRAE VKGVYQFCFS NEFSTFSHKT VYFDFQVGDE PPILPDMGNR VTALTQMESA CVTIHEALKT VIDSQTHYRL REAQDRARAE DLNSRVS.
Potential role in vesicular protein trafficking, mainly in the early secretory pathway. Contributes to the coupled localization of TMED2 and TMED10 in the cis-Golgi network. [UniProt].
Specifications
TMED3 | |
Polyclonal | |
Unconjugated | |
TMED3 | |
1200002G13Rik; AW546672; C15orf22; integral type I protein; membrane protein p24B; MGC133022; p24 family protein gamma-4; P24b; p24g4; p24gamma4; p26; testis tissue sperm-binding protein Li 48e; TMED3; transmembrane emp24 domain containing 3; transmembrane emp24 domain-containing protein 3; transmembrane emp24 domain-containing protein 3-like; transmembrane emp24 protein transport domain containing 3; transmembrane p24 trafficking protein 3; UNQ5357/PRO1078 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23423, 300888, 66111 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q6AY25, Q78IS1, Q9Y3Q3 | |
TMED3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 24-180 of human TMED3 (NP_031390.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction