Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMED4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987125
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7125 Supplier Bioss Supplier No. BS6208R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Involved in endoplasmic reticulum stress response. May play a role in the regulation of heat-shock response and apoptosis.

Specifications

Antigen TMED4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TMED4
Gene Accession No. Q7Z7H5
Gene Alias 1110014L17Rik; AI326346; Endoplasmic reticulum stress-response protein 25; endoplasmic reticulum stress-response protein 25 kDa; Ers25; GMP25iso; HNLF; p24 family protein alpha-3; p24a3; p24alpha3; p26; Putative NF-kappa-B-activating protein 156; putative NFkB activating protein HNLF; RGD1306319; Tmed4; Transmembrane emp24 domain-containing protein 4; transmembrane emp24 protein transport domain containing 4; transmembrane p24 trafficking protein 4
Gene Symbols TMED4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TMED4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222068
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.