Learn More
Invitrogen™ TMED5 Polyclonal Antibody

Description
Immunogen sequence: FELILDNMGE QAQEQEDWKK YITGTDILDM KLEDILESIN SIKSRLSKSG HIQTLL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
Specifications
Specifications
| Antigen | TMED5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TMED5 |
| Gene Accession No. | Q9Y3A6 |
| Gene Alias | CGI-100; p24 family protein gamma-2; p24g2; p24gamma2; p28; TMED5; Transmembrane emp24 domain-containing protein 5; transmembrane emp24 protein transport domain containing 5; transmembrane p24 trafficking protein 5; UNQ397/PRO733 |
| Gene Symbols | TMED5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.