Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMED9 Monoclonal Antibody (A9A8)
GREENER_CHOICE

Catalog No. FEMA550607
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550607 Supplier Invitrogen™ Supplier No. MA550607
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Hela cell lysate, THP-1 cell lysate, A549 cell lysate, MCF-7 cell lysate, PC-12 cell lysate, NIH/3T3 cell lysate, mouse liver tissue lysate, mouse lung tissue lysate, HepG2 cell lysate, LO2 cell lysate, human liver tissue lysate, rat liver tissue lysate, mouse liver tissue. Predicted band size: 27 kDa Subcellular Location: Endoplasmic reticulum membrane, Golgi apparatus, Endoplasmic reticulum-Golgi intermediate compartment membrane.

TMED9 (also named as GMP25, GP25L2 or p25) is a member of the EMP24/GP25L family. TMED9 probably forms hetero-oligomeric complexes with TMED7 (p27), TMED2 (p24), and TMED10 (p23). Located in the endoplasmic reticulum and the Golgi, TMED9 appears to be involved in vesicular protein trafficking, mainly in the early secretory pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMED9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone A9A8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TMED9
Gene Accession No. Q5I0E7, Q99KF1, Q9BVK6
Gene Alias 2400003B06Rik; glycoprotein 25L2; GMP25; Gp25l2; gp25L2 protein; HSGP25L2G; p24 family protein alpha-2; p24a2; p24alpha2; p25; RGD1307627; TMED9; Transmembrane emp24 domain-containing protein 9; transmembrane emp24 protein transport domain containing 9; transmembrane p24 trafficking protein 9
Gene Symbols TMED9
Host Species Mouse
Immunogen Recombinant protein within Human TMED9 aa 31-220/235.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361207, 54732, 67511
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.