Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMED9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110495 Supplier Invitrogen™ Supplier No. PA5110495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAVELGVLLV RPRPGTGLGR VMRTLLLVLW LATRGSALYF HIGETEKKCF IEEIPDETMV IGNYRTQLYD KQREEYQPAT PGLGMFVEVK DPEDKVILAR QYGSEGRFTF TSHTPGEHQI CLHSNSTKFS LFAGGMLRVH LDIQVGEHAN DYAEIAAKDK LSELQLRVRQ LVEQVEQIQK EQNYQRWREE RFRQTSESTN QRVLWWSILQ TLILVAIGVW QMRH.

TMED9 (also named as GMP25, GP25L2 or p25) is a member of the EMP24/GP25L family. TMED9 probably forms hetero-oligomeric complexes with TMED7 (p27), TMED2 (p24), and TMED10 (p23). Located in the endoplasmic reticulum and the Golgi, TMED9 appears to be involved in vesicular protein trafficking, mainly in the early secretory pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMED9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TMED9
Gene Accession No. Q5I0E7, Q99KF1, Q9BVK6
Gene Alias 2400003B06Rik; glycoprotein 25L2; GMP25; Gp25l2; gp25L2 protein; HSGP25L2G; p24 family protein alpha-2; p24a2; p24alpha2; p25; RGD1307627; TMED9; Transmembrane emp24 domain-containing protein 9; transmembrane emp24 protein transport domain containing 9; transmembrane p24 trafficking protein 9
Gene Symbols TMED9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-224 of human TMED9 (NP_059980.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361207, 54732, 67511
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.