Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEFF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569376
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The TMEFF1 gene encodes the protein transmembrane protein with ependymin domain 1, which is involved in cellular signaling and neural development. TMEFF1, characterized by its transmembrane and ependymin-related domains, plays a role in the modulation of cell-cell interactions and signaling pathways essential for brain development and function. This protein is expressed predominantly in the central nervous system, where it influences neuronal differentiation and synaptic formation, contributing to the establishment and maintenance of neural circuits. TMEFF1 has been implicated in neuroprotective functions, where it supports neuronal survival in response to stress or injury. Additionally, its expression has been studied in the context of cancer biology, particularly in brain tumors, where TMEFF1 might impact tumor growth and progression through its effects on cell signaling and adhesion. Research continues to explore the specific mechanisms and interactions of TMEFF1 to better understand its roles in neural health and disease, particularly in neurodevelopmental disorders and cancer.
Specifications
TMEFF1 | |
Polyclonal | |
Unconjugated | |
TMEFF1 | |
A830033E11; C9orf2; cancer/testis antigen family 120, member 1; CT120.1; H7365; H7365member 1; M7365; Nc1; Protein NC1; TMEFF1; Tomoregulin-1; Tr1; TR-1; transmembrane protein with EGF like and two follistatin like domains 1; transmembrane protein with EGF-like and one follistatin-like domain; transmembrane protein with EGF-like and two follistatin-like domains 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
8577 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IYR6 | |
TMEFF1 | |
synthetic peptide directed towards the middle region of human TMEFF1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction