Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEFF1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA562420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562420 Supplier Thermo Scientific Supplier No. PA562420

Rabbit Polyclonal Antibody

Immunogen sequence: DCPGGKGKSI NCSELNVRES Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 90%.

Tmeff1 may inhibit NODAL and BMP signaling during neural patterning.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEFF1
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEFF1
Gene Accession No. Q8IYR6
Gene Alias A830033E11; C9orf2; cancer/testis antigen family 120, member 1; CT120.1; H7365; H7365member 1; M7365; Nc1; Protein NC1; TMEFF1; Tomoregulin-1; Tr1; TR-1; transmembrane protein with EGF like and two follistatin like domains 1; transmembrane protein with EGF-like and one follistatin-like domain; transmembrane protein with EGF-like and two follistatin-like domains 1
Gene Symbols TMEFF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEFF1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8577
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.