Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEFF2 Recombinant Human Monoclonal Antibody (6G6)
GREENER_CHOICE

Catalog No. FEMA550086
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550086 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550086 Supplier Invitrogen™ Supplier No. MA550086
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

TMEFF2 Recombinant Monoclonal Antibody for ELISA

TMEFF2 may be a survival factor for hippocampal and mesencephalic neurons. The shedded form up-regulates cancer cell proliferation, probably by promoting ERK1/2 phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEFF2
Applications ELISA, Neutralization
Classification Recombinant Monoclonal
Clone 6G6
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene TMEFF2
Gene Accession No. Q9UIK5
Gene Alias 4832418D20Rik; 7630402F16Rik; cancer/testis antigen family 120, member 2; CT120.2; HPP1; Hyperplastic polyposis protein 1; MNCb-1026; TENB2; TMEFF2; tomoregulin; tomoregulin-2; TPEF; TR; TR-2; transmembrane protein TENB2; transmembrane protein with EGF like and two follistatin like domains 2; transmembrane protein with EGF-like and two follistatin-like domains; transmembrane protein with EGF-like and two follistatin-like domains 2; UNQ178/PRO204
Gene Symbols TMEFF2
Host Species Human
Immunogen Recombinant Human TMEFF2 protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23671
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.