Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM100 Antibody (OTI10G3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274545F
Description
TMEM100 Monoclonal specifically detects TMEM100 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TMEM100 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TMEM100 | |
Full length recombinant protein of human TMEM100 (NP_060756) produced in HEK293T cells. | |
0.1 mL | |
55273.0 | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI10G3 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
FLJ10970, FLJ37856, transmembrane protein 100 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction