Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM100 Antibody (OTI10G3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP148024
Description
TMEM100 Monoclonal specifically detects TMEM100 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TMEM100 | |
Monoclonal | |
0.4 mg/mL | |
Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
TMEM100 | |
Full length human recombinant protein of human TMEM100 (NP_060756) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
55273 | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI10G3 | |
Unconjugated | |
FLJ10970, FLJ37856, transmembrane protein 100 | |
Mouse | |
14.4 kDa | |
0.1 mL | |
Primary | |
This antibody is specific for Homo sapiens transmembrane protein 100 (TMEM100), transcript variant 2. | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction