Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM100 Monoclonal Antibody (OTI1F8)
GREENER_CHOICE

Catalog No. PIMA524941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524941 Supplier Invitrogen™ Supplier No. MA524941
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMEM100 Monoclonal Antibody for Western Blot, IHC (P)

Plays a role during embryonic arterial endothelium differentiation and vascular morphogenesis through the ACVRL1 receptor-dependent signaling pathway upon stimulation by bone morphogenetic proteins, such as GDF2/BMP9 and BMP10. Involved in the regulation of nociception, acting as a modulator of the interaction between TRPA1 and TRPV1, two molecular sensors and mediators of pain signals in dorsal root ganglia (DRG) neurons. Mechanistically, it weakens their interaction, thereby releasing the inhibition of TRPA1 by TRPV1 and increasing the single-channel open probability of the TRPA1-TRPV1 complex. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM100
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1F8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TMEM100
Gene Accession No. Q9NV29
Gene Alias EGK_08550; TMEM100; Transmembrane protein 100
Gene Symbols TMEM100
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human TMEM100 expression vector.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55273, 609661
Target Species Human, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.