Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM100 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548260
Description
Predicted to react with bovine based on sequence homology.
Plays a role during embryonic arterial endothelium differentiation and vascular morphogenesis through the ACVRL1 receptor-dependent signaling pathway upon stimulation by bone morphogenetic proteins, such as GDF2/BMP9 and BMP10. Involved in the regulation of nociception, acting as a modulator of the interaction between TRPA1 and TRPV1, two molecular sensors and mediators of pain signals in dorsal root ganglia (DRG) neurons. Mechanistically, it weakens their interaction, thereby releasing the inhibition of TRPA1 by TRPV1 and increasing the single-channel open probability of the TRPA1-TRPV1 complex. [UniProt].
Specifications
TMEM100 | |
Polyclonal | |
Unconjugated | |
TMEM100 | |
EGK_08550; TMEM100; Transmembrane protein 100 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
55273 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9NV29 | |
TMEM100 | |
KLH conjugated synthetic peptide between 93-122 amino acids from the C-terminal region of human TMEM100. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction