Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM100 Rabbit anti-Mouse, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN172125UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1721-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1721-25UG Supplier MilliporeSigma Supplier No. ABN172125UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-TMEM100, Cat. No. ABN1721, is a highly specific rabbit polyclonal antibody that targets Transmembrane protein 100 and has been tested in Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM100
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Transmembrane protein 100
Gene Symbols Tmem100
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminus region of murine Transmembrane protein 100 (TMEM100).
Molecular Weight of Antigen ∽17 kDa observed; 14.50 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects transmembrane protein 100 (TMEM100) in murine cells. It targets an epitope with the 19 amino acids from the N-terminal end.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.