Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM102 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555077
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555077 Supplier Invitrogen™ Supplier No. PA555077
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HAPLGPYARG PHYDAGFTLL VPMFSLDGTE LQLDLESCYA QVCLPEMVCG TPIREMWQDC LGPPVPGARD SIHRTESEES SKDWQSSVDQ PHSYVTEHE Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 62%.

Selectively involved in CSF2 deprivation-induced apoptosis via a mitochondria-dependent pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM102
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM102
Gene Accession No. Q8N9M5
Gene Alias 4632403M07Rik; Cbap; common beta-chain associated protein; common beta-chain-associated protein; LOW QUALITY PROTEIN: transmembrane protein 102; RGD1559450; TMEM102; Tmem102-ps; transmembrane protein 102; transmembrane protein 102, pseudogene
Gene Symbols TMEM102
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM102. Recombinant protein control fragment (Product #RP-93484).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284114
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.